Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257970.1 | NP_001244899.1 | 11 | 1742 | ||
NM_024110.4 | NP_077015.2 | 11 | 1742 | |||
NM_052819.2 | NP_438170.1 | 8 | 1023 | |||
NR_047566.1 | 12 | 1739 | ||||
XM_011525212.1 | XP_011523514.1 | 13 | 2263 | |||
XM_011525213.1 | XP_011523515.1 | 13 | 2005 | |||
XM_011525215.1 | XP_011523517.1 | 12 | 2065 | |||
XM_011525216.1 | XP_011523518.1 | 12 | 1765 | |||
XM_011525217.1 | XP_011523519.1 | 12 | 1821 | |||
XM_011525218.2 | XP_011523520.1 | 12 | 1804 | |||
XM_017025057.1 | XP_016880546.1 | 13 | 2260 | |||
XM_017025058.1 | XP_016880547.1 | 13 | 2263 | |||
GenBank mRNA | AF322642.1 | - | 11 | 1780 | ||
AK054723.1 | - | 3 | 458 | |||
AK091123.1 | - | 12 | 1697 | |||
AK296762.1 | - | 8 | 963 | |||
AK309299.1 | - | 7 | 1053 | |||
AX746790.1 | - | 12 | 1697 | |||
AY032927.1 | - | 10 | 1577 | |||
BC001326.1 | - | 8 | 1023 | |||
BC018142.1 | - | 11 | 1751 | 3847282 | ||
EU652409.1 | - | 10 | 1574 |