Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000496.2 | NP_000487.1 | 5 | 379 | ||
XM_006724141.3 | XP_006724204.1 | 5 | 1538 | |||
XM_011529900.2 | XP_011528202.1 | 6 | 1903 | |||
GenBank mRNA | BC069535.1 | - | 4 | 343 | 7262188 | |
CR456426.1 | - | 4 | 341 | |||
KR052782.1 | - | 5 | 373 | |||
KR052783.1 | - | 5 | 373 | |||
KR052784.1 | - | 5 | 375 | |||
KR052788.1 | - | 4 | 331 | |||
KR052789.1 | - | 4 | 322 | |||
KR052790.1 | - | 4 | 364 | |||
KR052791.1 | - | 3 | 279 | |||
L10035.1 | - | 4 | 343 |
Gene Symbol: CRYBB2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033733.1 | 4 | 471 | |||
NR_033734.1 | 3 | 352 | ||||
GenBank mRNA | BC037884.1 | - | 3 | 339 | ||
BC047380.1 | - | 4 | 474 |