Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024114.3 | NP_077019.2 | 6 | 1241 | ||
GenBank mRNA | AF521869.1 | - | 6 | 1248 | ||
BC001862.2 | - | 6 | 1241 | 3603763 |