Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032015.4 | NP_114404.1 | 1 | 1727 | ||
GenBank mRNA | AB055622.1 | - | 1 | 1415 | ||
AK075379.1 | - | 1 | 1485 | |||
AK122642.1 | - | 1 | 1700 | |||
BC000058.2 | - | 1 | 1496 | 3507662 | ||
BC007534.1 | - | 1 | 1467 | 3028875 | ||
BX647424.1 | - | 1 | 1700 | |||
BX648447.1 | - | 1 | 1703 |