Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024119.2 | NP_077024.2 | 9 | 1310 | ||
XM_017025059.1 | XP_016880548.1 | 8 | 1243 | |||
XM_017025060.1 | XP_016880549.1 | 9 | 1306 | |||
GenBank mRNA | AK021416.1 | - | 9 | 1310 | ||
AK097669.1 | - | 8 | 1142 | |||
AK225549.1 | - | 9 | 1310 | |||
AK296840.1 | - | 9 | 1269 | |||
AK300900.1 | - | 9 | 1240 | |||
BC014949.1 | - | 9 | 1247 | 4865798 |