Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039375.2 | NP_001034464.1 | 9 | 889 | ||
NM_024120.4 | NP_077025.2 | 10 | 973 | |||
NR_029377.1 | 11 | 1071 | ||||
XM_006723622.2 | XP_006723685.1 | 9 | 854 | |||
XM_006723623.2 | XP_006723686.1 | 9 | 986 | |||
XM_006723624.2 | XP_006723687.1 | 9 | 1000 | |||
XM_011529344.2 | XP_011527646.1 | 7 | 1328 | |||
XM_017028061.1 | XP_016883550.1 | 10 | 1085 | |||
GenBank mRNA | AK025977.1 | - | 4 | 399 | ||
AK091060.1 | - | 11 | 1048 | |||
AK289781.1 | - | 10 | 965 | |||
BC005984.1 | - | 10 | 783 | |||
BC073158.1 | - | 9 | 866 | 6053315 |