Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134671.2 | NP_001128143.1 | 4 | 647 | ||
NM_024295.5 | NP_077271.1 | 4 | 647 | |||
XM_006716657.1 | XP_006716720.1 | 3 | 131 | |||
XM_011517299.1 | XP_011515601.1 | 3 | 447 | |||
GenBank mRNA | AK023846.1 | - | 3 | 208 | ||
AK124086.1 | - | 3 | 208 | |||
AY358818.1 | - | 4 | 525 | |||
BC002457.2 | - | 4 | 476 | 3345230 |