Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011667.2 | NP_001011667.1 | 4 | 509 | ||
NM_001011668.2 | NP_001011668.1 | 4 | 465 | |||
NM_001011669.2 | NP_001011669.1 | 4 | 489 | |||
NM_001011670.2 | NP_001011670.1 | 3 | 361 | |||
NM_001011671.2 | NP_001011671.1 | 3 | 317 | |||
NM_001317858.1 | NP_001304787.1 | 4 | 489 | |||
NM_001317859.1 | NP_001304788.1 | 3 | 317 | |||
NM_024300.4 | NP_077276.2 | 4 | 445 | |||
NR_133934.1 | 4 | 509 | ||||
NR_133935.1 | 4 | 509 | ||||
NR_133936.1 | 4 | 465 | ||||
NR_133937.1 | 3 | 357 | ||||
GenBank mRNA | AK095922.1 | - | 3 | 166 | ||
AK098285.1 | - | 2 | 2968 | |||
AK225982.1 | - | 4 | 304 | |||
AK290088.1 | - | 4 | 294 | |||
AY070434.1 | - | 4 | 337 | |||
BC002546.2 | - | 4 | 284 | 3139026 | ||
BI911444.1 | - | 4 | 320 | |||
BM695222.1 | - | 3 | 201 | |||
BX473986.1 | - | 4 | 347 | |||
CN256918.1 | - | 3 | 147 | |||
DA367603.1 | - | 4 | 358 | |||
DA431755.1 | - | 4 | 358 | |||
DA988698.1 | - | 4 | 314 |