Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004076.4 | NP_004067.1 | 5 | 468 | ||
XM_011529902.2 | XP_011528204.1 | 5 | 1006 | |||
GenBank mRNA | BC069479.1 | - | 5 | 438 | 7262237 | |
BC102021.1 | - | 5 | 425 | 40029651 | ||
BC102022.1 | - | 5 | 425 | 40029650 | ||
BC107482.1 | - | 5 | 425 | 40029652 | ||
BM711382.1 | - | 5 | 468 | |||
CR456427.1 | - | 4 | 419 |