Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024308.3 | NP_077284.2 | 3 | 653 | ||
XM_005257658.2 | XP_005257715.1 | 3 | 658 | |||
XM_011525233.2 | XP_011523535.1 | 2 | 393 | |||
GenBank mRNA | AB209247.1 | - | 1 | 1947 | ||
AK026196.1 | - | 3 | 444 | |||
AK315735.1 | - | 3 | 574 | |||
AY358712.1 | - | 3 | 550 | |||
BC002731.2 | - | 3 | 539 | 3631177 | ||
CR457356.1 | - | 2 | 202 | |||
LC110386.1 | - | 3 | 439 |