Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024310.4 | NP_077286.3 | 2 | 1099 | ||
XM_005259256.3 | XP_005259313.1 | 2 | 1387 | |||
XM_011527309.2 | XP_011525611.1 | 2 | 2373 | |||
GenBank mRNA | AK027758.1 | - | 2 | 1058 | ||
AY037145.2 | - | 2 | 1463 | |||
BC002744.2 | - | 2 | 1053 | 3628796 | ||
BC018876.2 | - | 2 | 1053 |