Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR15L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024320.3 | NP_077296.1 | 2 | 323 | ||
XM_005257663.4 | XP_005257720.1 | 2 | 353 | |||
XM_005257664.2 | XP_005257721.1 | 1 | 109 | |||
XM_005257665.1 | XP_005257722.1 | 1 | 109 | |||
XM_005257666.4 | XP_005257723.1 | 3 | 434 | |||
GenBank mRNA | BC002865.2 | - | 2 | 190 | 3939938 | |
BM829856.1 | - | 2 | 323 |