Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319113.1 | NP_001306042.1 | 2 | 358 | ||
NM_024321.4 | NP_077297.2 | 2 | 358 | |||
XM_005259257.2 | XP_005259314.1 | 2 | 353 | |||
GenBank mRNA | AK301669.1 | - | 2 | 283 | ||
BC002868.2 | - | 2 | 297 | 3941786 | ||
BC004204.2 | - | 2 | 296 | 3532802 | ||
BC031682.1 | - | 2 | 325 | 5163682 |