Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199803.2 | NP_001186732.1 | 3 | 600 | ||
NM_001322101.1 | NP_001309030.1 | 4 | 714 | |||
NM_024322.3 | NP_077298.1 | 4 | 733 | |||
NR_136182.1 | 3 | 619 | ||||
NR_136183.1 | 3 | 619 | ||||
NR_136184.1 | 3 | 600 | ||||
GenBank mRNA | AK027859.1 | - | 3 | 334 | ||
AK315483.1 | - | 4 | 359 | |||
AL514456.3 | - | 3 | 293 | |||
BC002870.1 | - | 4 | 390 | 3942844 | ||
BP366489.1 | - | 4 | 386 | |||
BX406097.2 | - | 4 | 389 | |||
CF993857.1 | - | 3 | 628 | |||
CN263048.1 | - | 3 | 256 | |||
CX872478.1 | - | 3 | 237 |