Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199803.2 | NP_001186732.1 | 6 | 1148 | ||
NM_001322101.1 | NP_001309030.1 | 7 | 1262 | |||
NM_024322.3 | NP_077298.1 | 7 | 1281 | |||
NR_136182.1 | 6 | 1066 | ||||
NR_136183.1 | 6 | 1167 | ||||
NR_136184.1 | 6 | 1047 | ||||
GenBank mRNA | AK315483.1 | - | 7 | 907 | ||
AL514456.3 | - | 6 | 841 | |||
BC002870.1 | - | 7 | 938 | 3942844 | ||
CX872478.1 | - | 6 | 684 |