Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145347.1 | NP_001138819.1 | 3 | 903 | ||
NM_024327.2 | NP_077303.1 | 3 | 939 | |||
GenBank mRNA | AK026353.1 | - | 3 | 939 |