Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126339.3 | NP_001119811.1 | 2 | 738 | ||
NM_024328.5 | NP_077304.1 | 1 | 1137 | |||
GenBank mRNA | AF432862.1 | - | 1 | 406 | ||
AK057691.1 | - | 1 | 844 | |||
AK098604.1 | - | 2 | 527 | |||
BC002984.1 | - | 1 | 831 | 3535451 | ||
BI522644.1 | - | 2 | 542 | |||
BM925597.1 | - | 2 | 725 | |||
BX161435.1 | - | 1 | 1097 |