Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126339.3 | NP_001119811.1 | 2 | 633 | ||
NM_001256062.2 | NP_001242991.1 | 1 | 1032 | |||
NM_001256321.2 | NP_001243250.1 | 2 | 633 | |||
NM_001256322.2 | NP_001243251.1 | 1 | 1032 | |||
NM_001256323.2 | NP_001243252.1 | 2 | 633 | |||
NM_024328.5 | NP_077304.1 | 1 | 1032 | |||
GenBank mRNA | AF432862.1 | - | 1 | 301 | ||
AK057691.1 | - | 1 | 739 | |||
AK098604.1 | - | 2 | 422 | |||
BC002984.1 | - | 1 | 726 | 3535451 | ||
BI522644.1 | - | 2 | 437 | |||
BM925597.1 | - | 2 | 620 | |||
BX161435.1 | - | 1 | 992 | |||
BX378775.2 | - | 1 | 720 | |||
DA952722.1 | - | 2 | 347 | |||
DB122722.1 | - | 2 | 344 |