Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTPAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039199.2 | NP_001034288.1 | 4 | 785 | ||
NM_001261839.1 | NP_001248768.1 | 4 | 683 | |||
NM_024331.4 | NP_077307.2 | 5 | 809 | |||
XM_011529043.2 | XP_011527345.1 | 5 | 921 | |||
XM_011529044.2 | XP_011527346.1 | 5 | 1536 | |||
XM_017028063.1 | XP_016883552.1 | 5 | 698 | |||
GenBank mRNA | AK125126.1 | - | 1 | 815 | ||
AK307236.1 | - | 2 | 501 | |||
AK314045.1 | - | 4 | 744 | |||
BC003071.2 | - | 5 | 788 | 2989837 | ||
BQ646722.1 | - | 4 | 649 | |||
CR936621.1 | - | 4 | 739 |