Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004297.2 | NP_001004297.2 | 4 | 950 | ||
XM_011540147.2 | XP_011538449.1 | 3 | 1417 | |||
XM_017016632.1 | XP_016872121.1 | 4 | 4336 | |||
XM_017016633.1 | XP_016872122.1 | 3 | 4478 | |||
XM_017016634.1 | XP_016872123.1 | 3 | 4723 | |||
GenBank mRNA | AK131555.1 | - | 4 | 950 | ||
BC104853.1 | - | 2 | 753 | 8143856 | ||
BC104855.1 | - | 2 | 753 | 8143858 | ||
BC143622.1 | - | 2 | 753 | 9052135 |