Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030763.2 | NP_110390.1 | 6 | 559 | ||
XM_006724688.3 | XP_006724751.1 | 6 | 509 | |||
XM_017029839.1 | XP_016885328.1 | 4 | 389 | |||
XM_017029840.1 | XP_016885329.1 | 4 | 383 | |||
GenBank mRNA | AF250329.1 | - | 5 | 298 | ||
AK094058.1 | - | 6 | 559 | |||
AK315792.1 | - | 5 | 352 | |||
BC005342.1 | - | 5 | 319 | 3838879 | ||
DA745749.1 | - | 6 | 528 |