Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030764.3 | NP_110391.2 | 9 | 1393 | ||
NR_125358.1 | 6 | 541 | ||||
XM_011509974.2 | XP_011508276.1 | 9 | 1728 | |||
XM_011509975.2 | XP_011508277.1 | 9 | 1730 | |||
XM_011509976.2 | XP_011508278.1 | 9 | 1729 | |||
XM_017002316.1 | XP_016857805.1 | 9 | 1728 | |||
XM_017002317.1 | XP_016857806.1 | 9 | 1531 | |||
XM_017002318.1 | XP_016857807.1 | 8 | 1451 | |||
XM_017002319.1 | XP_016857808.1 | 8 | 1443 | |||
GenBank mRNA | AF319438.1 | - | 4 | 878 | ||
AF390037.1 | - | 4 | 716 | |||
AF459633.1 | - | 9 | 1381 | |||
AL833194.1 | - | 4 | 3252 | |||
AY043465.1 | - | 9 | 1380 | |||
AY358130.1 | - | 9 | 1383 | |||
BC069185.1 | - | 6 | 545 | |||
BC129836.1 | - | 4 | 718 | 40115446 |