Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184779.1 | NP_001171708.1 | 6, 7 | 963 | ||
NM_001184780.1 | NP_001171709.1 | 6, 7 | 1090 | |||
NM_024505.3 | NP_078781.3 | 6, 7 | 1047 | |||
NR_033671.2 | 7, 8 | 1278 | ||||
NR_033672.1 | 7, 8 | 1194 | ||||
GenBank mRNA | AF317889.1 | - | 3, 4 | 478 | ||
AF325189.1 | - | 7, 8 | 1169 | |||
AF325190.1 | - | 7, 8 | 1253 | |||
AF353088.1 | - | 6, 7 | 942 | |||
AF353089.1 | - | 6, 7 | 1026 | |||
AK026011.1 | - | 3, 4 | 532 | |||
AK074058.1 | - | 4, 5 | 716 | |||
AK074071.1 | - | 5, 6 | 4165 | |||
AK314689.1 | - | 6, 7 | 1026 | |||
AY166601.1 | - | 3, 4 | 406 | |||
BC125097.1 | - | 6, 7 | 956 | 40118286 | ||
BC125098.1 | - | 7, 8 | 1060 | 40118288 | ||
DQ413001.1 | - | 3, 4 | 406 |