Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024513.3 | NP_078789.2 | 8 | 2012 | ||
XM_006713333.3 | XP_006713396.1 | 9 | 3177 | |||
XM_006713334.3 | XP_006713397.1 | 8 | 3093 | |||
XM_011534111.2 | XP_011532413.1 | 9 | 2090 | |||
GenBank mRNA | AJ292348.2 | - | 8 | 1996 | ||
AL832358.1 | - | 8 | 2000 | |||
BC101468.1 | - | 7 | 1882 | 8068974 | ||
BC101470.1 | - | 7 | 1882 | 8068976 | ||
BC143368.1 | - | 7 | 1882 | 9051873 |