Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130042.1 | NP_001123514.1 | 9 | 1256 | ||
NM_001130043.1 | NP_001123515.1 | 7 | 1115 | |||
NM_001134759.1 | NP_001128231.1 | 6 | 929 | |||
NM_001889.3 | NP_001880.2 | 8 | 1217 | |||
XM_005270491.4 | XP_005270548.1 | 8 | 730 | |||
XM_011540747.1 | XP_011539049.1 | 8 | 836 | |||
XM_017000367.1 | XP_016855856.1 | 8 | 789 | |||
XM_017000368.1 | XP_016855857.1 | 7 | 740 | |||
GenBank mRNA | AB209714.1 | - | 9 | 888 | ||
AK123469.1 | - | 1 | 1297 | |||
AK128794.1 | - | 8 | 822 | |||
AK223150.1 | - | 8 | 849 | |||
AK223201.1 | - | 8 | 849 | |||
AK314813.1 | - | 7 | 755 | |||
BC039578.1 | - | 8 | 839 | 6067125 | ||
BC070058.1 | - | 9 | 898 | 5267616 | ||
BX647883.1 | - | 6 | 540 | |||
BX649061.1 | - | 1 | 1850 | |||
L13278.1 | - | 7 | 761 |