Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005484.1 | NP_001005484.1 | 1 | 113 |
Gene Symbol: OR4F17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005240.1 | NP_001005240.1 | 1 | 113 | ||
XM_005259648.1 | XP_005259705.1 | 2 | 256 | |||
GenBank mRNA | AY972817.1 | - | 2 | 262 | ||
AY972818.1 | - | 1 | 179 | |||
BC136848.1 | - | 1 | 149 | 9020839 | ||
BC136867.1 | - | 1 | 149 | 9020858 |
Gene Symbol: OR4F4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004195.2 | NP_001004195.2 | 1 | 113 | ||
GenBank mRNA | AY972819.1 | - | 1 | 179 | ||
BC136907.1 | - | 1 | 149 | 9020898 | ||
BC136908.1 | - | 1 | 149 | 9020899 |