Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004063.2 | NP_001004063.2 | 1 | 464 | ||
XM_011537153.2 | XP_011535455.1 | 3 | 576 | |||
GenBank mRNA | BC151145.1 | - | 1 | 464 | 9020994 |