Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004077.2 | NP_004068.2 | 7 | 835 | ||
GenBank mRNA | AF047042.1 | - | 7 | 645 | ||
AF053631.1 | - | 7 | 810 | |||
AF147410.1 | - | 5 | 535 | |||
AK074956.1 | - | 7 | 683 | |||
AK095856.1 | - | 7 | 670 | |||
AK293263.1 | - | 5 | 620 | |||
AK296249.1 | - | 8 | 815 | |||
AK307625.1 | - | 3 | 252 | |||
BC000105.1 | - | 5 | 475 | |||
BC010106.1 | - | 7 | 763 | 3542491 | ||
BC072016.1 | - | 7 | 678 | 6484456 | ||
BT007414.1 | - | 1 | 39 | |||
BX640838.1 | - | 8 | 892 |