Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM65A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193522.1 | NP_001180451.1 | 13 | 1717 | ||
NM_001193523.1 | NP_001180452.1 | 13 | 1830 | |||
NM_001193524.1 | NP_001180453.1 | 13 | 1743 | |||
NM_024519.3 | NP_078795.2 | 13 | 1708 | |||
XM_011523321.1 | XP_011521623.1 | 13 | 2522 | |||
XM_011523322.1 | XP_011521624.1 | 13 | 1717 | |||
XM_011523324.2 | XP_011521626.1 | 13 | 1777 | |||
XM_011523325.1 | XP_011521627.1 | 13 | 1732 | |||
XM_011523326.1 | XP_011521628.1 | 13 | 1931 | |||
XM_017023663.1 | XP_016879152.1 | 13 | 1717 | |||
XM_017023664.1 | XP_016879153.1 | 12 | 1603 | |||
GenBank mRNA | AB067517.1 | - | 12 | 1567 | ||
AK023787.1 | - | 8 | 1200 | |||
AK097087.1 | - | 12 | 1958 | |||
AK127792.1 | - | 13 | 1717 | |||
AK293748.1 | - | 13 | 1740 | |||
AK295677.1 | - | 13 | 1674 | |||
AK302868.1 | - | 3 | 867 | |||
AK307467.1 | - | 13 | 1661 | |||
AL834312.1 | - | 3 | 773 | |||
BC054512.1 | - | 3 | 750 | |||
BC098587.1 | - | 7 | 1145 |