Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024522.2 | NP_078798.2 | 6 | 896 | ||
XM_017002320.1 | XP_016857809.1 | 5 | 490 | |||
GenBank mRNA | AK022712.1 | - | 5 | 468 | ||
AK298874.1 | - | 4 | 387 | |||
AK299507.1 | - | 6 | 568 | |||
AL832547.1 | - | 3 | 161 | |||
BC096707.1 | - | 5 | 454 | 40022279 | ||
BC098173.1 | - | 5 | 454 | 40022282 | ||
BC098249.1 | - | 5 | 454 | 40022276 | ||
BC131518.1 | - | 5 | 454 | 40022275 |