Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004077.2 | NP_004068.2 | 7 | 912 | ||
GenBank mRNA | AF047042.1 | - | 7 | 722 | ||
AF053631.1 | - | 7 | 887 | |||
AK074956.1 | - | 7 | 760 | |||
AK095856.1 | - | 7 | 747 | |||
AK293263.1 | - | 5 | 697 | |||
AK296249.1 | - | 8 | 892 | |||
AK307625.1 | - | 3 | 329 | |||
BC000105.1 | - | 5 | 552 | |||
BC010106.1 | - | 7 | 840 | 3542491 | ||
BC072016.1 | - | 7 | 755 | 6484456 | ||
BT007414.1 | - | 1 | 116 | |||
BX640838.1 | - | 8 | 969 | |||
DQ403132.1 | - | 1 | 56 |