Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122835.2 | NP_001116307.2 | 16 | 1953 | ||
NM_024525.4 | NP_078801.3 | 18 | 2115 | |||
XM_005273260.1 | XP_005273317.1 | 18 | 2112 | |||
XM_005273261.2 | XP_005273318.1 | 17 | 2022 | |||
XM_005273262.1 | XP_005273319.1 | 16 | 1956 | |||
XM_005273264.2 | XP_005273321.1 | 16 | 1887 | |||
XM_006711814.2 | XP_006711877.1 | 14 | 1728 | |||
XM_006711815.2 | XP_006711878.1 | 11 | 3122 | |||
XM_011544275.2 | XP_011542577.1 | 15 | 1794 | |||
XM_011544276.2 | XP_011542578.1 | 11 | 1358 | |||
XM_017002321.1 | XP_016857810.1 | 17 | 2019 | |||
XM_017002322.1 | XP_016857811.1 | 16 | 1884 | |||
XM_017002323.1 | XP_016857812.1 | 14 | 1690 | |||
XM_017002324.1 | XP_016857813.1 | 14 | 1725 | |||
XM_017002325.1 | XP_016857814.1 | 18 | 5317 | |||
XM_017002326.1 | XP_016857815.1 | 13 | 4697 | |||
GenBank mRNA | AK026237.1 | - | 5 | 422 | ||
AK074365.1 | - | 9 | 1023 | |||
AK075386.1 | - | 18 | 2081 | |||
AK075561.1 | - | 16 | 1932 | |||
AK309725.1 | - | 14 | 1771 | |||
AL832286.1 | - | 18 | 1850 | |||
AL834453.1 | - | 9 | 1081 | |||
BC042683.1 | - | 9 | 999 |