Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122835.2 | NP_001116307.2 | 21 | 2416 | ||
NM_024525.4 | NP_078801.3 | 23 | 2578 | |||
XM_005273260.1 | XP_005273317.1 | 23 | 2575 | |||
XM_005273261.2 | XP_005273318.1 | 22 | 2485 | |||
XM_005273262.1 | XP_005273319.1 | 21 | 2419 | |||
XM_005273264.2 | XP_005273321.1 | 21 | 2350 | |||
XM_006711814.2 | XP_006711877.1 | 18 | 2341 | |||
XM_006711815.2 | XP_006711878.1 | 16 | 3585 | |||
XM_011544275.2 | XP_011542577.1 | 20 | 2257 | |||
XM_011544276.2 | XP_011542578.1 | 16 | 1821 | |||
XM_017002321.1 | XP_016857810.1 | 22 | 2482 | |||
XM_017002322.1 | XP_016857811.1 | 21 | 2347 | |||
XM_017002323.1 | XP_016857812.1 | 19 | 2153 | |||
XM_017002324.1 | XP_016857813.1 | 19 | 2188 | |||
XM_017002325.1 | XP_016857814.1 | 23 | 5780 | |||
XM_017002326.1 | XP_016857815.1 | 18 | 5160 | |||
GenBank mRNA | AK026237.1 | - | 10 | 885 | ||
AK074365.1 | - | 14 | 1486 | |||
AK075386.1 | - | 23 | 2544 | |||
AK075561.1 | - | 21 | 2395 | |||
AL832286.1 | - | 23 | 2313 | |||
AL834453.1 | - | 14 | 1544 | |||
BC042683.1 | - | 14 | 1462 |