Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004077.2 | NP_004068.2 | 7 | 803 | ||
GenBank mRNA | AF053631.1 | - | 7 | 778 | ||
AF147410.1 | - | 5 | 503 | |||
AK074956.1 | - | 7 | 651 | |||
AK095856.1 | - | 7 | 638 | |||
AK293263.1 | - | 5 | 588 | |||
AK296249.1 | - | 8 | 783 | |||
AK307625.1 | - | 3 | 220 | |||
BC000105.1 | - | 5 | 443 | |||
BC010106.1 | - | 7 | 731 | 3542491 | ||
BC072016.1 | - | 7 | 646 | 6484456 | ||
BX640838.1 | - | 8 | 860 |