Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201472.1 | NP_001188401.1 | 21 | 2454 | ||
NM_001201473.1 | NP_001188402.1 | 19 | 2253 | |||
NM_024535.4 | NP_078811.3 | 21 | 2508 | |||
GenBank mRNA | AK025674.1 | - | 21 | 2117 | ||
AK094596.1 | - | 21 | 2127 | |||
AK097238.1 | - | 21 | 1974 | |||
AK126176.1 | - | 7 | 1280 | |||
AK294045.1 | - | 21 | 2089 | |||
AK296723.1 | - | 21 | 2086 | |||
AK296807.1 | - | 19 | 1919 | |||
BC117289.1 | - | 21 | 2127 | 40125840 | ||
BC117291.1 | - | 21 | 2127 | 40125842 |
Gene Symbol: CORO7-PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201479.1 | NP_001188408.1 | 21 | 2508 | ||
GenBank mRNA | AL833954.1 | - | 23 | 2583 |