Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195731.1 | NP_001182660.1 | 2 | 445 | ||
NM_024536.5 | NP_078812.2 | 2 | 801 | |||
XM_011511838.2 | XP_011510140.1 | 1 | 251 | |||
GenBank mRNA | AB086063.1 | - | 2 | 635 | ||
AB095813.1 | - | 2 | 810 | |||
AK302124.1 | - | 2 | 445 | |||
AY358403.1 | - | 2 | 801 | |||
BC021223.2 | - | 2 | 801 | 4128192 | ||
BC023531.2 | - | 2 | 752 | 4123041 | ||
DC341343.1 | - | 2 | 398 |