Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195731.1 | NP_001182660.1 | 4 | 2602 | ||
NM_024536.5 | NP_078812.2 | 4 | 2958 | |||
XM_011511838.2 | XP_011510140.1 | 3 | 2489 | |||
GenBank mRNA | AB095813.1 | - | 4 | 2967 | ||
AK026331.1 | - | 1 | 1382 | |||
AY358403.1 | - | 4 | 2958 | |||
BC008878.2 | - | 1 | 1510 | |||
BC021223.2 | - | 4 | 2958 | 4128192 | ||
BC023531.2 | - | 4 | 2909 | 4123041 |