Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024537.3 | NP_078813.1 | 13, 14 | 1500 | ||
XM_006719953.2 | XP_006720016.1 | 13, 14 | 1580 | |||
XM_011521116.1 | XP_011519418.1 | 14, 15 | 1342 | |||
XM_017020741.1 | XP_016876230.1 | 12, 13 | 1228 | |||
GenBank mRNA | AB062436.1 | - | 13, 14 | 1417 | ||
AK022180.1 | - | 13, 14 | 1436 | |||
AK091992.1 | - | 5, 6 | 1917 | |||
BC007220.1 | - | 13, 14 | 1423 | 2822557 |