Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024540.3 | NP_078816.2 | 5 | 541 | ||
NM_145729.2 | NP_663781.1 | 5 | 582 | |||
XM_011509981.2 | XP_011508283.1 | 5 | 652 | |||
XM_011509982.2 | XP_011508284.1 | 5 | 550 | |||
GenBank mRNA | AK222500.1 | - | 5 | 539 | ||
AK223066.1 | - | 5 | 539 | |||
BC012440.1 | - | 5 | 523 | 3863359 | ||
BC016700.1 | - | 5 | 541 | 4108596 | ||
BG706700.1 | - | 5 | 546 | |||
CR457329.1 | - | 4 | 402 |