Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024541.2 | NP_078817.2 | 24 | 1931 | ||
XM_006717969.1 | XP_006718032.1 | 22 | 1719 | |||
XM_011540151.2 | XP_011538453.1 | 24 | 1976 | |||
XM_011540152.2 | XP_011538454.1 | 23 | 1863 | |||
XM_011540153.2 | XP_011538455.1 | 23 | 1827 | |||
XM_011540154.2 | XP_011538456.1 | 22 | 1761 | |||
XM_011540156.2 | XP_011538458.1 | 17 | 1895 | |||
XM_011540157.2 | XP_011538459.1 | 23 | 1880 | |||
XM_017016635.1 | XP_016872124.1 | 24 | 1929 | |||
XM_017016636.1 | XP_016872125.1 | 24 | 1929 | |||
GenBank mRNA | AK023176.1 | - | 7 | 560 | ||
AK097972.1 | - | 1 | 294 | |||
BC110511.1 | - | 6 | 484 |