Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSE1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256135.1 | NP_001243064.1 | 17 | 1890 | ||
NM_001316.3 | NP_001307.2 | 18 | 2058 | |||
NR_045796.1 | 15 | 1573 | ||||
XM_011528599.2 | XP_011526901.1 | 18 | 1992 | |||
GenBank mRNA | AF053641.1 | - | 18 | 1992 | ||
AK098618.1 | - | 4 | 338 | |||
AK225610.1 | - | 10 | 931 | |||
AK297270.1 | - | 14 | 1359 | |||
AK297314.1 | - | 17 | 1891 | |||
AK298477.1 | - | 11 | 1054 | |||
AK299549.1 | - | 9 | 859 | |||
AK300589.1 | - | 15 | 1508 | |||
AK307965.1 | - | 11 | 1004 | |||
AK312306.1 | - | 18 | 1989 | |||
BC108309.1 | - | 18 | 2004 | 4540265 | ||
BC109313.2 | - | 18 | 1971 | 40036129 | ||
BC109314.2 | - | 18 | 1971 | 40036130 | ||
EF426455.1 | - | 16 | 1701 | |||
U33286.1 | - | 18 | 1992 |