Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145928.1 | NP_001139400.1 | 16 | 2410 | ||
NM_024545.3 | NP_078821.2 | 15 | 2305 | |||
XM_005263767.3 | XP_005263824.1 | 16 | 2381 | |||
XM_005263768.3 | XP_005263825.1 | 16 | 2360 | |||
XM_005263769.3 | XP_005263826.1 | 16 | 2281 | |||
XM_005263770.3 | XP_005263827.1 | 16 | 2357 | |||
XM_006712748.3 | XP_006712811.1 | 16 | 2567 | |||
XM_006712749.3 | XP_006712812.1 | 15 | 2462 | |||
XM_017004903.1 | XP_016860392.1 | 16 | 2357 | |||
XM_017004904.1 | XP_016860393.1 | 15 | 2255 | |||
XM_017004905.1 | XP_016860394.1 | 15 | 2252 | |||
GenBank mRNA | AK022823.1 | - | 6 | 927 | ||
AK092234.1 | - | 9 | 1363 | |||
AK315185.1 | - | 16 | 2410 | |||
AL136833.1 | - | 15 | 2303 | |||
AY220791.1 | - | 15 | 2173 | |||
BC017453.1 | - | 8 | 1110 | |||
BC117255.1 | - | 15 | 2271 | 40125806 | ||
BC143898.1 | - | 16 | 2376 | 9052416 |