Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024548.3 | NP_078824.2 | 9 | 1256 | ||
XM_006713743.3 | XP_006713806.1 | 8 | 1647 | |||
XM_011513127.1 | XP_011511429.1 | 9 | 1213 | |||
GenBank mRNA | AK093100.1 | - | 6 | 787 | ||
AK309806.1 | - | 8 | 1155 | |||
AL833269.1 | - | 9 | 1206 | |||
BC041085.1 | - | 9 | 1199 | 5528397 |