Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303401.1 | NP_001290330.1 | 11 | 2188 | ||
NM_024548.3 | NP_078824.2 | 11 | 2365 | |||
XM_006713743.3 | XP_006713806.1 | 10 | 2756 | |||
XM_011513127.1 | XP_011511429.1 | 11 | 2322 | |||
XM_017007178.1 | XP_016862667.1 | 10 | 2581 | |||
GenBank mRNA | AK294784.1 | - | 11 | 2126 | ||
AL833269.1 | - | 11 | 2315 | |||
BC041085.1 | - | 11 | 2308 | 5528397 |