Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNJL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308173.1 | NP_001295102.1 | 5 | 1013 | ||
NM_024565.6 | NP_078841.3 | 6 | 1157 | |||
NR_131769.1 | 6 | 1015 | ||||
XM_006714917.3 | XP_006714980.1 | 5 | 1592 | |||
XM_011534646.2 | XP_011532948.1 | 5 | 895 | |||
XM_011534647.2 | XP_011532949.1 | 3 | 947 | |||
XM_017009846.1 | XP_016865335.1 | 6 | 1240 | |||
XM_017009847.1 | XP_016865336.1 | 4 | 792 | |||
XM_017009848.1 | XP_016865337.1 | 4 | 1152 | |||
XM_017009849.1 | XP_016865338.1 | 3 | 529 | |||
GenBank mRNA | AK024228.1 | - | 1 | 132 | ||
AK131381.1 | - | 6 | 1015 | |||
AK294000.1 | - | 5 | 889 | |||
AK302823.1 | - | 5 | 1013 | |||
BC013353.2 | - | 2 | 193 | |||
BC035871.1 | - | 6 | 1123 | 5245637 |