Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HMBOX1 SIRNA Assays ›
Gene Symbol:
HMBOX1
Gene Name:
homeobox containing 1
Gene Aliases:
HNF1LA, HOT1, PBHNF
Chromosome Location:
Chr.8: 28890370 - 29064939 on Build GRCh38
UniGene ID:
Hs.563560
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35928
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HMBOX1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135726.2
NP_001129198.1
3
409
NM_001324382.1
NP_001311311.1
3
269
NM_001324383.1
NP_001311312.1
3
409
NM_001324384.1
NP_001311313.1
3
412
NM_001324385.1
NP_001311314.1
3
409
NM_001324386.1
NP_001311315.1
3
409
NM_001324387.1
NP_001311316.1
3
409
NM_001324388.1
NP_001311317.1
3
269
NM_001324389.1
NP_001311318.1
3
409
NM_001324390.1
NP_001311319.1
3
412
NM_001324391.1
NP_001311320.1
3
409
NM_001324392.1
NP_001311321.1
3
412
NM_001324393.1
NP_001311322.1
4
525
NM_001324394.1
NP_001311323.1
3
409
NM_001324395.1
NP_001311324.1
3
409
NM_024567.4
NP_078843.2
4
443
NR_136757.1
3
409
NR_136758.1
3
409
XM_005273635.1
XP_005273692.1
3
385
XM_005273641.4
XP_005273698.1
3
385
XM_006716398.2
XP_006716461.1
3
385
XM_011544642.2
XP_011542944.1
3
385
XM_017013823.1
XP_016869312.1
3
417
XM_017013824.1
XP_016869313.1
3
207
XM_017013825.1
XP_016869314.1
3
385
GenBank mRNA
AK290683.1
-
3
385
AK295320.1
-
2
48
AK310959.1
-
3
375
AY522342.1
-
3
154
BC069242.1
-
4
443
5928365
DC341559.1
-
4
276
DC358620.1
-
3
229
DC395744.1
-
3
270
DQ269478.1
-
3
154
HY077398.1
-
4
441
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
RNA-dependent DNA biosynthetic process
transcription, DNA-templated
positive regulation of telomere maintenance via telomerase
positive regulation of chromatin binding
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of telomerase activity
double-stranded telomeric DNA binding
telomerase activity
protein binding
protein complex binding
telomeric DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0