Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024571.3 | NP_078847.1 | 4 | 478 | ||
GenBank mRNA | BC001381.1 | - | 4 | 1300 | 3051476 | |
BC009179.1 | - | 4 | 429 | 3946937 | ||
Z84714.1 | - | 4 | 392 |