Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324266.1 | NP_001311195.1 | 5 | 1029 | ||
NM_024576.4 | NP_078852.3 | 5 | 1026 | |||
XM_011536117.2 | XP_011534419.1 | 5 | 962 | |||
XM_011536118.2 | XP_011534420.1 | 5 | 811 | |||
XM_017011303.1 | XP_016866792.1 | 5 | 591 | |||
XM_017011304.1 | XP_016866793.1 | 5 | 588 | |||
XM_017011305.1 | XP_016866794.1 | 5 | 484 | |||
XM_017011306.1 | XP_016866795.1 | 5 | 1514 | |||
GenBank mRNA | AK024732.1 | - | 2 | 89 | ||
BC018688.1 | - | 5 | 661 | 4717736 | ||
BC110999.1 | - | 5 | 665 | 5585901 | ||
BX420584.2 | - | 5 | 695 |