Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301039.1 | NP_001287968.1 | 5 | 355 | ||
NM_001301040.1 | NP_001287969.1 | 5 | 355 | |||
NM_001301041.1 | NP_001287970.1 | 3 | 141 | |||
NM_001301042.1 | NP_001287971.1 | 4 | 225 | |||
NM_024579.3 | NP_078855.2 | 4 | 244 | |||
XM_011509984.2 | XP_011508286.1 | 4 | 271 | |||
GenBank mRNA | AK026874.1 | - | 4 | 229 | ||
AX770997.1 | - | 4 | 242 | |||
AX770998.1 | - | 5 | 269 | |||
AX770999.1 | - | 4 | 244 | |||
BC017761.1 | - | 4 | 229 | 4617936 | ||
DA358536.1 | - | 3 | 152 |