Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288705.1 | NP_001275634.1 | 5 | 978 | ||
NM_005211.3 | NP_005202.2 | 6 | 1057 | |||
NR_109969.1 | 5 | 978 | ||||
GenBank mRNA | AB208819.1 | - | 5 | 928 | ||
AK294467.1 | - | 5 | 893 | |||
AK304491.1 | - | 5 | 759 | |||
BC047521.1 | - | 5 | 960 | 4819527 | ||
BC129939.1 | - | 5 | 898 | |||
BX648599.1 | - | 4 | 848 | |||
CD620053.1 | - | 1 | 50 | |||
EU826593.1 | - | 5 | 765 | |||
X03663.1 | - | 6 | 1065 |